siRNA Details
| Gene Symbol: Dpf3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001267626.1 | NP_001254555.1 | 9 | 1016 | ||
| NM_058212.2 | NP_478119.1 | 9 | 1013 | |||
| GenBank mRNA | AF362750.1 | - | 9 | 985 | ||
| AK012970.1 | - | 9 | 1021 | |||
| BC048572.1 | - | 9 | 896 | 6705809 | ||
| FJ151218.1 | - | 9 | 988 | |||
More Information
Other Species Targeted
This siRNA also targets the following transcripts in Rat: NM_001191818, XM_008764748.

Dpf3
Genomic Map