siRNA Details
| Gene Symbol: 2210011C24Rik | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001291292.1 | NP_001278221.1 | 1 | 329 | ||
| GenBank mRNA | AI847924.1 | - | 1 | 20 | ||
| AK008705.1 | - | 1 | 51 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

2210011C24Rik| Gene Symbol: 2210011C24Rik | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001291292.1 | NP_001278221.1 | 1 | 329 | ||
| GenBank mRNA | AI847924.1 | - | 1 | 20 | ||
| AK008705.1 | - | 1 | 51 | |||