siRNA Details
| Gene Symbol: Med23 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001166416.1 | NP_001159888.1 | 21 | 2671 | ||
| NM_027347.3 | NP_081623.3 | 20 | 2653 | |||
| GenBank mRNA | AK028545.1 | - | 21 | 2684 | ||
| AK129312.1 | - | 20 | 2586 | |||
| BC005508.1 | - | 4 | 516 | |||
| BC040398.1 | - | 20 | 2587 | |||
| BC050916.1 | - | 20 | 2592 | 6405418 | ||

Med23
Genomic Map