siRNA Details
| Gene Symbol: Ppil3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001285826.1 | NP_001272755.1 | 6 | 1056 | ||
| NM_001285827.1 | NP_001272756.1 | 6 | 721 | |||
| NM_027351.3 | NP_081627.1 | 6 | 535 | |||
| NM_027374.3 | NP_081650.2 | 6 | 721 | |||
| GenBank mRNA | AF315803.1 | - | 6 | 669 | ||
| AK010207.1 | - | 6 | 539 | |||
| AK011000.1 | - | 6 | 490 | |||
| BB687332.1 | - | 1 | 27 | |||
| BC060993.1 | - | 6 | 503 | 6591278 | ||
| BC061645.1 | - | 6 | 999 | 5032617 | ||
| DV043111.1 | - | 6 | 757 | |||

Ppil3
Genomic Map