siRNA Details
| Gene Symbol: Cd2bp2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001285905.1 | NP_001272834.1 | 4 | 483 | ||
| NM_001285906.1 | NP_001272835.1 | 4 | 497 | |||
| NM_001285907.1 | NP_001272836.1 | 4 | 625 | |||
| NM_027353.4 | NP_081629.1 | 4 | 477 | |||
| GenBank mRNA | AK010582.1 | - | 4 | 362 | ||
| AK030393.1 | - | 4 | 359 | |||
| AK031694.1 | - | 4 | 391 | |||
| AK034689.1 | - | 4 | 401 | |||
| AK154346.1 | - | 4 | 362 | |||
| AK164102.1 | - | 3 | 341 | |||
| AK167249.1 | - | 4 | 423 | |||
| AK173122.1 | - | 3 | 319 | |||
| BC006920.1 | - | 4 | 393 | 3601764 | ||
| BC079575.1 | - | 3 | 551 | |||
| BM944739.1 | - | 4 | 407 | |||

Cd2bp2
Genomic Map