siRNA Details
| Gene Symbol: Bhlhb9 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001098222.1 | NP_001091692.1 | 5 | 1528 | ||
| NM_198161.2 | NP_937804.1 | 5 | 1538 | |||
| GenBank mRNA | AK012577.1 | - | 1 | 21 | ||
| BC059871.1 | - | 1 | 893 | 6809364 | ||

Bhlhb9
Genomic Map