siRNA Details
| Gene Symbol: Gtf3c5 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001290484.1 | NP_001277413.1 | 8 | 1444 | ||
| NM_148928.3 | NP_683730.1 | 8 | 1444 | |||
| GenBank mRNA | AK045419.1 | - | 8 | 1036 | ||
| AK147014.1 | - | 8 | 1422 | |||
| BC023847.1 | - | 1 | 55 | |||
| BC027055.1 | - | 8 | 1411 | 5360397 | ||
| BC027247.1 | - | 8 | 1354 | 2631223 | ||
| BC039171.1 | - | 8 | 1365 | 4500799 | ||

Gtf3c5
Genomic Map