siRNA Details
| Gene Symbol: 2010107E04Rik | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_027360.2 | NP_081636.1 | 4 | 217 | ||
| GenBank mRNA | AK008333.1 | - | 4 | 218 | ||
| BC055712.1 | - | 4 | 196 | 6408438 | ||
| BC059719.1 | - | 4 | 205 | 6774233 | ||

2010107E04Rik
Genomic Map