siRNA Details
| Gene Symbol: Gcc2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_027375.2 | NP_081651.2 | 21 | 4795 | ||
| GenBank mRNA | AB093232.1 | - | 21 | 4798 | ||
| AK148283.1 | - | 16 | 2010 | |||
| BC023811.1 | - | 16 | 2179 | |||
| BC034077.1 | - | 10 | 1433 | |||
| BC065697.1 | - | 9 | 1190 | |||
| BC139007.1 | - | 21 | 4791 | 8862029 | ||
| BC145495.1 | - | 21 | 4864 | |||

Gcc2
Genomic Map