siRNA Details
| Gene Symbol: Gcc2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_027375.2 | NP_081651.2 | 6 | 535 | ||
| GenBank mRNA | AB093232.1 | - | 6 | 537 | ||
| AK011206.1 | - | 6 | 553 | |||
| AK045701.1 | - | 6 | 542 | |||
| BC027339.1 | - | 6 | 517 | |||
| BC139007.1 | - | 6 | 531 | 8862029 | ||
| BC145495.1 | - | 6 | 604 | |||
| EF410116.1 | - | 536 | ||||

Gcc2
Genomic Map