siRNA Details
| Gene Symbol: Arl16 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_197995.2 | NP_932112.2 | 5 | 546 | ||
| GenBank mRNA | AK011161.1 | - | 3 | 226 | ||
| AK038349.1 | - | 5 | 418 | |||
| AK140077.1 | - | 3 | 712 | |||
| BC009089.1 | - | 3 | 1103 | |||
| BC139260.1 | - | 5 | 417 | 8862282 | ||
| BC139262.1 | - | 5 | 417 | 8862284 | ||

Arl16
Genomic Map