siRNA Details
| Gene Symbol: Mospd1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001290514.1 | NP_001277443.1 | 5 | 810 | ||
| NM_027409.5 | NP_081685.1 | 5 | 813 | |||
| GenBank mRNA | AK007533.1 | - | 5 | 746 | ||
| AK088275.1 | - | 5 | 747 | |||
| AK139795.1 | - | 5 | 754 | |||
| AK203602.1 | - | 1 | 61 | |||
| BC018329.1 | - | 5 | 711 | 3962906 | ||
| BC085100.1 | - | 5 | 718 | 6391017 | ||
| BC100442.1 | - | 5 | 698 | 30501889 | ||
| DV651027.1 | - | 2 | 180 | |||
More Information
Other Species Targeted
This siRNA also targets the following transcripts in Rat: NM_001014107, XM_006257632, XM_006257633, XM_006257631, XM_006251400, and others.

Mospd1
Genomic Map