siRNA Details
| Gene Symbol: Mospd1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001290514.1 | NP_001277443.1 | 5, 6 | 858 | ||
| NM_027409.5 | NP_081685.1 | 5, 6 | 861 | |||
| GenBank mRNA | AK007533.1 | - | 5, 6 | 794 | ||
| AK203602.1 | - | 1, 2 | 109 | |||
| BC018329.1 | - | 5, 6 | 759 | 3962906 | ||
| BC085100.1 | - | 5, 6 | 766 | 6391017 | ||
| BC100442.1 | - | 5, 6 | 746 | 30501889 | ||
| DV651027.1 | - | 2, 3 | 228 | |||

Mospd1
Genomic Map