siRNA Details
| Gene Symbol: Tmem70 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_026392.1 | NP_080668.1 | 3 | 459 | ||
| NM_027415.2 | NP_081691.2 | 3 | 456 | |||
| GenBank mRNA | AK003839.1 | - | 3 | 441 | ||
| AK005075.1 | - | 3 | 441 | |||
| AK008955.1 | - | 3 | 425 | |||
| AK151280.1 | - | 3 | 431 | |||
| AK167069.1 | - | 3 | 382 | |||
| AK169031.1 | - | 3 | 389 | |||
| BC011320.1 | - | 3 | 385 | 3979025 | ||

Tmem70
Genomic Map