siRNA Details
| Gene Symbol: Tspan15 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_197996.2 | NP_932113.2 | 2 | 247 | ||
| GenBank mRNA | AK012471.1 | - | 2 | 246 | ||
| AK172169.1 | - | 2 | 257 | |||
| BC003872.1 | - | 2 | 196 | 3492667 | ||
| CN455615.1 | - | 2 | 136 | |||
| CX206586.1 | - | 2 | 247 | |||

Tspan15
Genomic Map