siRNA Details
| Gene Symbol: Mier2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_027422.2 | NP_081698.2 | 6 | 566 | ||
| GenBank mRNA | AK154672.1 | - | 6 | 583 | ||
| AK173128.1 | - | 5 | 722 | |||
| BC067013.1 | - | 6 | 566 | 6810181 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Mier2| Gene Symbol: Mier2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_027422.2 | NP_081698.2 | 6 | 566 | ||
| GenBank mRNA | AK154672.1 | - | 6 | 583 | ||
| AK173128.1 | - | 5 | 722 | |||
| BC067013.1 | - | 6 | 566 | 6810181 | ||