siRNA Details
Gene Symbol: Mpc2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_027430.2 | NP_081706.1 | 1 | 249 | ||
GenBank mRNA | AK002233.2 | - | 1 | 198 | ||
AK008044.1 | - | 1 | 59 | |||
AK011889.1 | - | 1 | 254 | |||
AK147721.1 | - | 822 | ||||
AK147792.1 | - | 823 | ||||
AK151522.1 | - | 1 | 236 | |||
AK163436.1 | - | 1 | 234 | |||
BC018324.1 | - | 1 | 95 | 3501265 |
