siRNA Details
| Gene Symbol: Mpc2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_027430.2 | NP_081706.1 | 3, 4 | 436 | ||
| GenBank mRNA | AK002233.2 | - | 3, 4 | 385 | ||
| AK011889.1 | - | 3, 4 | 441 | |||
| AK151522.1 | - | 3, 4 | 423 | |||
| BC018324.1 | - | 3, 4 | 282 | 3501265 | ||

Mpc2
Genomic Map