siRNA Details
| Gene Symbol: Mpc2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_027430.2 | NP_081706.1 | 5 | 563 | ||
| GenBank mRNA | AK011889.1 | - | 5 | 568 | ||
| AK151522.1 | - | 5 | 550 | |||
| BC018324.1 | - | 5 | 409 | 3501265 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Mpc2| Gene Symbol: Mpc2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_027430.2 | NP_081706.1 | 5 | 563 | ||
| GenBank mRNA | AK011889.1 | - | 5 | 568 | ||
| AK151522.1 | - | 5 | 550 | |||
| BC018324.1 | - | 5 | 409 | 3501265 | ||