siRNA Details
| Gene Symbol: Atad2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_027435.2 | NP_081711.2 | 19 | 2598 | ||
| GenBank mRNA | AK029867.1 | - | 12 | 1745 | ||
| AK134319.1 | - | 6 | 974 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Atad2| Gene Symbol: Atad2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_027435.2 | NP_081711.2 | 19 | 2598 | ||
| GenBank mRNA | AK029867.1 | - | 12 | 1745 | ||
| AK134319.1 | - | 6 | 974 | |||