siRNA Details
Gene Symbol: Atad2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_027435.2 | NP_081711.2 | 4 | 617 | ||
GenBank mRNA | AK012079.1 | - | 4 | 605 | ||
AK032877.1 | - | 4 | 618 |

Please enter the information below and press OK to send your cart to Core Services for purchase.
Gene Symbol: Atad2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_027435.2 | NP_081711.2 | 4 | 617 | ||
GenBank mRNA | AK012079.1 | - | 4 | 605 | ||
AK032877.1 | - | 4 | 618 |