siRNA Details
Gene Symbol: 5730403I07Rik | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | AK017485.1 | - | 2, 3 | 234 | ||
AK161639.1 | - | 2, 3 | 130 | |||
BC048919.1 | - | 2, 3 | 132 |

Please enter the information below and press OK to send your cart to Core Services for purchase.
Gene Symbol: 5730403I07Rik | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | AK017485.1 | - | 2, 3 | 234 | ||
AK161639.1 | - | 2, 3 | 130 | |||
BC048919.1 | - | 2, 3 | 132 |