siRNA Details
| Gene Symbol: 5730403I07Rik | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| GenBank mRNA | AK017485.1 | - | 3 | 272 | ||
| AK161639.1 | - | 3 | 168 | |||
| BC048919.1 | - | 3 | 170 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

5730403I07Rik| Gene Symbol: 5730403I07Rik | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| GenBank mRNA | AK017485.1 | - | 3 | 272 | ||
| AK161639.1 | - | 3 | 168 | |||
| BC048919.1 | - | 3 | 170 | |||