siRNA Details
| Gene Symbol: Tmem242 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_027457.4 | NP_081733.3 | 2 | 167 | ||
| GenBank mRNA | AK003559.2 | - | 2 | 166 | ||
| AK009851.1 | - | 2 | 150 | |||
| AK017626.1 | - | 2 | 164 | |||
| AK043276.1 | - | 2 | 135 | |||
| AK088025.1 | - | 2 | 147 | |||
| AK155326.1 | - | 2 | 146 | |||
| AK158107.1 | - | 2 | 146 | |||

Tmem242
Genomic Map