siRNA Details
| Gene Symbol: Tln2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001081242.2 | NP_001074711.2 | 20 | 2489 | ||
| GenBank mRNA | AF467081.1 | - | 18 | 2218 | ||
| AK165499.1 | - | 14 | 1661 | |||
| BC046480.1 | - | 12 | 1461 | |||
| BC079896.1 | - | 11 | 1450 | 30544295 | ||

Tln2
Genomic Map