siRNA Details
| Gene Symbol: Arhgap27 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_183288.3 | NP_899111.2 | 3 | 1218 | ||
| GenBank mRNA | AK017636.1 | - | 3 | 910 | ||
| BC138292.1 | - | 1 | 674 | 8861311 | ||
| BC138293.1 | - | 1 | 674 | 8861312 | ||
| Gene Symbol: Arhgap27os2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| GenBank mRNA | AK052773.1 | - | 2 | 548 | ||

Arhgap27
Genomic Map