siRNA Details
| Gene Symbol: Cpne3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_027769.2 | NP_082045.1 | 8 | 787 | ||
| GenBank mRNA | AK017651.1 | - | 8 | 792 | ||
| AK036664.1 | - | 8 | 766 | |||
| AK146324.1 | - | 8 | 764 | |||
| AK220348.1 | - | 8 | 758 | |||
| BC036983.1 | - | 8 | 714 | |||
| BC090632.1 | - | 7 | 688 | 30620886 | ||

Cpne3
Genomic Map