siRNA Details
| Gene Symbol: Tcerg1l | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_183289.3 | NP_899112.2 | 1, 2 | 414 | ||
| GenBank mRNA | AK017700.1 | - | 1, 2 | 414 | ||
| BC107234.1 | - | 1, 2 | 29 | |||
| BC107235.1 | - | 1, 2 | 29 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Tcerg1l| Gene Symbol: Tcerg1l | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_183289.3 | NP_899112.2 | 1, 2 | 414 | ||
| GenBank mRNA | AK017700.1 | - | 1, 2 | 414 | ||
| BC107234.1 | - | 1, 2 | 29 | |||
| BC107235.1 | - | 1, 2 | 29 | |||