siRNA Details
| Gene Symbol: Ipo5 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_023579.4 | NP_076068.1 | 6 | 678 | ||
| GenBank mRNA | AF294327.1 | - | 6 | 629 | ||
| AK017701.1 | - | 6 | 637 | |||
| AK049422.1 | - | 6 | 650 | |||
| AK053681.1 | - | 6 | 643 | |||
| AK079134.1 | - | 6 | 648 | |||
| AK167303.1 | - | 6 | 640 | |||
| AK169664.1 | - | 6 | 646 | |||
| BC052392.1 | - | 6 | 621 | 5719784 | ||
| BC054814.1 | - | 6 | 851 | 6842146 | ||
| CB524964.1 | - | 6 | 637 | |||

Ipo5
Genomic Map