siRNA Details
| Gene Symbol: Zdhhc24 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001168516.1 | NP_001161988.1 | 2 | 165 | ||
| NM_001168517.1 | NP_001161989.1 | 2 | 212 | |||
| NM_027476.3 | NP_081752.2 | 2 | 458 | |||
| GenBank mRNA | AK017737.1 | - | 2 | 423 | ||
| AK028164.1 | - | 2 | 421 | |||
| AK043923.1 | - | 2 | 415 | |||
| AK169337.1 | - | 2 | 417 | |||
| AK170706.1 | - | 2 | 442 | |||
| AK171568.1 | - | 2 | 442 | |||
| CA559443.1 | - | 2 | 165 | |||
| CD354313.1 | - | 2 | 121 | |||

Zdhhc24
Genomic Map