siRNA Details
| Gene Symbol: 5730508B09Rik | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_027482.3 | NP_081758.1 | 2 | 509 | ||
| GenBank mRNA | AK012470.1 | - | 2 | 480 | ||
| AK017758.1 | - | 2 | 472 | |||
| AK162420.1 | - | 2 | 486 | |||
| BC039568.1 | - | 2 | 236 | |||
| BC116791.1 | - | 2 | 456 | 40126110 | ||
| BC116793.1 | - | 2 | 456 | 40126112 | ||
| BC144728.1 | - | 3 | 487 | |||

5730508B09Rik
Genomic Map