siRNA Details
| Gene Symbol: Vcpip1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| GenBank mRNA | AK030104.1 | - | 2 | 2972 | ||
| AK122563.1 | - | 2 | 2985 | |||
| BC059209.1 | - | 2 | 2991 | 6406239 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Vcpip1| Gene Symbol: Vcpip1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| GenBank mRNA | AK030104.1 | - | 2 | 2972 | ||
| AK122563.1 | - | 2 | 2985 | |||
| BC059209.1 | - | 2 | 2991 | 6406239 | ||