siRNA Details
| Gene Symbol: Utp20 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_175158.3 | NP_780367.2 | 61, 62 | 8376 | ||
| GenBank mRNA | AB052760.1 | - | 19, 20 | 2576 | ||
| AK014430.1 | - | 7, 8 | 1004 | |||
| AK047081.1 | - | 2427 | ||||
| BC048955.1 | - | 31, 32 | 4241 | |||

Utp20
Genomic Map