siRNA Details
| Gene Symbol: Adgra2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_133911.1 | NP_598672.1 | 9 | 1423 | ||
| GenBank mRNA | AK014455.1 | - | 9 | 1394 | ||
| AK028929.1 | - | 9 | 1362 | |||
| BC020317.2 | - | 4 | 556 | |||
| BC052391.1 | - | 9 | 1423 | |||
More Information
Set Membership: |
|

Adgra2
Genomic Map