siRNA Details
| Gene Symbol: Hmha1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001142701.1 | NP_001136173.1 | 16 | 2099 | ||
| NM_027521.3 | NP_081797.1 | 15 | 1651 | |||
| GenBank mRNA | AK018130.1 | - | 3 | 166 | ||
| AK044860.1 | - | 15 | 1613 | |||
| AK134029.1 | - | 9 | 1073 | |||
| AK151423.1 | - | 16 | 2043 | |||
| AK159194.1 | - | 16 | 2100 | |||
| AK159474.1 | - | 15 | 2144 | |||
| AK161766.1 | - | 8 | 1159 | |||
| AK170262.1 | - | 16 | 2085 | |||
| AK171304.1 | - | 16 | 2085 | |||
| BC053750.1 | - | 15 | 1651 | 6333971 | ||

Hmha1
Genomic Map