siRNA Details
| Gene Symbol: Hmha1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001142701.1 | NP_001136173.1 | 21 | 3031 | ||
| NM_027521.3 | NP_081797.1 | 20 | 2583 | |||
| GenBank mRNA | AK018130.1 | - | 8 | 1098 | ||
| AK044860.1 | - | 20 | 2545 | |||
| AK134029.1 | - | 14 | 2005 | |||
| AK151423.1 | - | 21 | 2975 | |||
| AK159194.1 | - | 21 | 3032 | |||
| AK159474.1 | - | 20 | 3076 | |||
| AK161766.1 | - | 13 | 2091 | |||
| AK170262.1 | - | 21 | 3017 | |||
| AK171304.1 | - | 21 | 3017 | |||
| BC039802.1 | - | 2 | 240 | |||
| BC053750.1 | - | 20 | 2583 | 6333971 | ||

Hmha1
Genomic Map