siRNA Details
| Gene Symbol: Angptl6 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_145154.2 | NP_660136.1 | 2 | 233 | ||
| GenBank mRNA | AB054065.1 | - | 2 | 233 | ||
| BC025904.1 | - | 2 | 211 | 5049765 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Angptl6| Gene Symbol: Angptl6 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_145154.2 | NP_660136.1 | 2 | 233 | ||
| GenBank mRNA | AB054065.1 | - | 2 | 233 | ||
| BC025904.1 | - | 2 | 211 | 5049765 | ||