siRNA Details
| Gene Symbol: Rasgef1a | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_027526.1 | NP_081802.1 | 2 | 260 | ||
| GenBank mRNA | AK018120.1 | - | 1 | 91 | ||
| AK158756.1 | - | 3 | 286 | |||
| BC138284.1 | - | 2 | 226 | 8861303 | ||
| BC145232.1 | - | 2 | 226 | 9053784 | ||
| BC145752.1 | - | 2 | 226 | 40131055 | ||
| BY120855.1 | - | 2 | 265 | |||
| BY256830.1 | - | 2 | 208 | |||

Rasgef1a
Genomic Map