siRNA Details
| Gene Symbol: Nos1ap | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001109985.1 | NP_001103455.1 | 2, 3 | 660 | ||
| NM_027528.2 | NP_081804.1 | 2, 3 | 477 | |||
| GenBank mRNA | AK018149.1 | - | 2, 3 | 660 | ||
| AK045124.1 | - | 2, 3 | 614 | |||
| AK053892.1 | - | 2, 3 | 661 | |||
| AK138471.1 | - | 2, 3 | 478 | |||
| AK162919.1 | - | 2, 3 | 478 | |||

Nos1ap
Genomic Map