siRNA Details
| Gene Symbol: Prdm5 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_027547.2 | NP_081823.2 | 10 | 1262 | ||
| GenBank mRNA | AK014501.1 | - | 10 | 1186 | ||
| AK018310.1 | - | 6 | 648 | |||
| BC138901.1 | - | 10 | 1236 | 8861923 | ||
| BC138902.1 | - | 10 | 1236 | 8861924 | ||

Prdm5
Genomic Map