siRNA Details
| Gene Symbol: Zdhhc1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_175160.3 | NP_780369.1 | 7, 8 | 939 | ||
| GenBank mRNA | AK038593.1 | - | 7, 8 | 941 | ||
| BC026570.1 | - | 3, 4 | 520 | 4984252 | ||
| BC119378.1 | - | 4, 5 | 678 | 8734127 | ||
| BC144878.1 | - | 4, 5 | 678 | 9053426 | ||

Zdhhc1
Genomic Map