siRNA Details
| Gene Symbol: Pgrmc2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_027558.1 | NP_081834.1 | 1, 2 | 459 | ||
| GenBank mRNA | AK014543.1 | - | 1, 2 | 358 | ||
| AK140064.1 | - | 1, 2 | 439 | |||
| AX775753.1 | - | 1, 2 | 415 | |||
| BC044759.2 | - | 1, 2 | 400 | |||
| CD363842.1 | - | 1, 2 | 458 | |||

Pgrmc2
Genomic Map