siRNA Details
| Gene Symbol: Pgrmc2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_027558.1 | NP_081834.1 | 2 | 569 | ||
| GenBank mRNA | AK014543.1 | - | 2 | 468 | ||
| AK184727.1 | - | 1 | 119 | |||
| AX775753.1 | - | 2 | 525 | |||
| BC044759.2 | - | 2 | 510 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Pgrmc2| Gene Symbol: Pgrmc2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_027558.1 | NP_081834.1 | 2 | 569 | ||
| GenBank mRNA | AK014543.1 | - | 2 | 468 | ||
| AK184727.1 | - | 1 | 119 | |||
| AX775753.1 | - | 2 | 525 | |||
| BC044759.2 | - | 2 | 510 | |||