siRNA Details
Gene Symbol: Cfap43 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_027559.2 | NP_081835.2 | 2 | 331 | ||
GenBank mRNA | AK014581.1 | - | 349 | |||
AK015217.1 | - | 2 | 353 | |||
AK015497.1 | - | 2 | 332 |

Please enter the information below and press OK to send your cart to Core Services for purchase.
Gene Symbol: Cfap43 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_027559.2 | NP_081835.2 | 2 | 331 | ||
GenBank mRNA | AK014581.1 | - | 349 | |||
AK015217.1 | - | 2 | 353 | |||
AK015497.1 | - | 2 | 332 |