siRNA Details
| Gene Symbol: Cfap43 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_027559.2 | NP_081835.2 | 4 | 621 | ||
| GenBank mRNA | AK014581.1 | - | 639 | |||
| AK015217.1 | - | 4 | 643 | |||
| AK015497.1 | - | 4 | 622 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Cfap43| Gene Symbol: Cfap43 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_027559.2 | NP_081835.2 | 4 | 621 | ||
| GenBank mRNA | AK014581.1 | - | 639 | |||
| AK015217.1 | - | 4 | 643 | |||
| AK015497.1 | - | 4 | 622 | |||