siRNA Details
| Gene Symbol: Hmgxb4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_178017.1 | NP_821136.1 | 9 | 1665 | ||
| GenBank mRNA | AK088758.1 | - | 5 | 491 | ||
| BC023167.1 | - | 5 | 407 | |||
| BC048188.1 | - | 10 | 1780 | |||
| BC049800.1 | - | 9 | 1665 | 6509446 | ||

Hmgxb4
Genomic Map