siRNA Details
| Gene Symbol: Hmgxb4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_178017.1 | NP_821136.1 | 5 | 552 | ||
| GenBank mRNA | AK014640.1 | - | 5 | 572 | ||
| AK156670.1 | - | 5 | 583 | |||
| BC023721.1 | - | 5 | 557 | |||
| BC048188.1 | - | 6 | 667 | |||
| BC049800.1 | - | 5 | 552 | 6509446 | ||

Hmgxb4
Genomic Map