siRNA Details
| Gene Symbol: Cnbd2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_027585.2 | NP_081861.2 | 7 | 982 | ||
| GenBank mRNA | AK014912.1 | - | 7 | 1005 | ||
| AK162372.1 | - | 7 | 863 | |||
| BC027204.1 | - | 7 | 786 | 4037010 | ||
| BC075727.1 | - | 4 | 566 | |||
| BC140978.1 | - | 7 | 965 | 9055590 | ||
| JN629039.1 | - | 7 | 922 | |||
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
|---|---|---|---|---|---|---|
| GenBankmRNA | AK139677.1 | - | 162 | Chr. - - [-] on Build | ||

Cnbd2
Genomic Map