siRNA Details
| Gene Symbol: Dmrtc1a | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001038616.2 | NP_001033705.2 | 4 | 435 | ||
| NM_027591.3 | NP_081867.1 | 6 | 533 | |||
| NM_029378.2 | NP_083654.1 | 4 | 347 | |||
| GenBank mRNA | AK006548.1 | - | 4 | 450 | ||
| AK014934.1 | - | 6 | 534 | |||
| BC147175.1 | - | 6 | 505 | 9056223 | ||
| BC147176.1 | - | 6 | 505 | 9056216 | ||
| DQ316772.1 | - | 6 | 489 | |||
| DQ316773.1 | - | 4 | 347 | |||
| DQ316774.1 | - | 4 | 347 | |||

Dmrtc1a
Genomic Map