siRNA Details
| Gene Symbol: 4921524L21Rik | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_027598.1 | NP_081874.1 | 7 | 675 | ||
| GenBank mRNA | AK014961.1 | - | 7 | 676 | ||
| BC049627.1 | - | 5 | 520 | 6706381 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

4921524L21Rik| Gene Symbol: 4921524L21Rik | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_027598.1 | NP_081874.1 | 7 | 675 | ||
| GenBank mRNA | AK014961.1 | - | 7 | 676 | ||
| BC049627.1 | - | 5 | 520 | 6706381 | ||